Would you like to log in again?
CLP–Semi-Volatiles Acid Surrogate Standard (3/90 SOW), Restek
Supplier: Restek
31073 31083 31003
RK31083EA 179.45 USD InStock
RK31083 RK31073 RK31003
CLP–Semi-Volatiles Acid Surrogate Standard (3/90 SOW), Restek
Standards Environmental Analysis Standards
Standards are packaged in methanol at the stated concentration in an ampoule.
  • Mixture contains 2-Chlorophenol-d4, 2-Fluorophenol, Phenol-d6, and 2,4,6-Tribromophenol
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty