Would you like to log in again?
SV Calibration Mix #6 (18 Components), Restek
Supplier: Restek
31012
RK31012EA 156.17 USD InStock
RK31012
SV Calibration Mix #6 (18 Components), Restek
Standards Organic Standards
Contains: Aldrin (309-00-2); α-BHC (319-84-6); β-BHC (319-85-7); δ-BHC (319-86-8); γ-BHC (Lindane) (58-89-9); 4,4'-DDD (72-54-8); 4,4'-DDE (72-55-9); 4,4'-DDT (50-29-3); Dieldrin (60-57-1); Endosulfan I (959-98-8); Endosulfan II (33213-65-9); Endosulfan sulfate (1031-07-8); Endrin (72-20-8); Endrin aldehyde (7421-93-4); Endrin ketone (53494-70-5); Heptachlor (76-44-8); Heptachlor epoxide (isomer B) (1024-57-3); Methoxychlor (72-43-5)
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty