Would you like to log in again?
TCLP Acid Mix (6 Components), Restek
Supplier: Restek
31027
RK31027EA 37.83 USD InStock
RK31027
TCLP Acid Mix (6 Components), Restek
Standards Organic Standards
TCLP acid mix contains 2-methylphenol (o-cresol), 3-methylphenol (m-cresol), 4-methylphenol (p-cresol), pentachlorophenol, 2,4,5-trichlorophenol, 2,4,6-trichlorophenol.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty