Would you like to log in again?
Environmental standards, Semi-Volatiles (SV) Internal Standard Mix, Restek
Supplier: Restek
31006-15 31006-25 31006 31206 31206-25 31206-15
RK31206EA 77.6 USD InStock
RK31206 75834-596 75834-632 75835-616 75834-634 RK31006
Environmental standards, Semi-Volatiles (SV) Internal Standard Mix, Restek
Standards Environmental Analysis Standards
Standards are packaged in methylene chloride in a 1 mL ampoule.
  • High purity for consistent results
  • Free quality assurance data package available online
  • Highest concentrations commercially available.
Mixture contains Acenaphthene-d10, Chrysene-d12, 1,4-Dichlorobenzene-d4, Naphthalene-d8, Perylene-d12, and Phenanthrene
US Environmental Protection Agency (EPA) Method 8270D outlines the analysis of semivolatile organic pollutants in solid waste, soil, water, and air matrices, using gas chromatography/mass spectrometry (GC/MS). Update IVA of the third edition of SW-846—Test Methods for Evaluating Solid Waste, Physical/Chemical Methods—includes EPA Method 8270D, in which there were no major revisions from EPA Method 8270C.
Delivery information: Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty