Would you like to log in again?
Kerosene Composite Standard, Restek
Supplier: Restek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
31256 31257
RK31256EA 49.47 USD InStock
RK31256 RK31257
Kerosene Composite Standard, Restek
Standards Petroleum Industry Standards
  • Kerosene composite standards are comprised of three separate sources blended in equal portions.
  • Kerosene standards may exhibit lot-to-lot variation.
Contains: Kerosene composite (84742-81-0)
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty