Would you like to log in again?
610 PAH Calibration Mix A (16 components), Restek
Supplier: Restek
31264
RK31264EA 62.08 USD InStock
RK31264
610 PAH Calibration Mix A (16 components), Restek
Standards Environmental Analysis Standards
610 PAH calibration mix A contains acenaphthene (1000 μg/ml), acenaphthylene (1000 μg/ml),anthracene (1000 μg/ml), benz(a)anthracene (500 μg/ml), benzo(a)pyrene (500 μg/ml), benzo(b)fluoranthene (500 μg/ml)
benzo(k)fluoranthene (500 μg/ml), benzo(ghi)perylene (500 μg/ml), chrysene (500 μg/ml), dibenz(a,h)anthracene (500 μg/ml), fluoranthene (500 μg/ml), fluorene (1000 μg/ml), indeno(1,2,3-cd)pyrene (500 μg/ml), naphthalene (1000 μg/ml), phenanthrene (500 μg/ml), pyrene ( 500 μg/ml).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty