Would you like to log in again?
MA EPH Matrix Spike Mix (10 components), Restek
Supplier: Restek
31460
RK31460EA 41.71 USD InStock
RK31460
MA EPH Matrix Spike Mix (10 components), Restek
Standards Organic Standards
Contains: n-nonane, C9, n-tetradecane, C14, n-nonadecane, C19, n-eicosane, C20, n-octacosane, C28, acenaphthene, anthracene, chrysene, naphthalene , pyrene.
Contains: n-nonane, C9, n-tetradecane, C14, n-nonadecane, C19, n-eicosane, C20, n-octacosane, C28, acenaphthene, anthracene, chrysene, naphthalene , pyrene.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty