Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Appendix IX Mix #2 (32 components), Restek
Supplier: Restek
31806
RK31806EA 92.15 USD InStock
RK31806
Appendix IX Mix #2 (32 components), Restek
Standards Environmental Analysis Standards
Contains: Acetophenone (98-86-2); Aramite (140-57-8); Atrazine (1912-24-9); Benzaldehyde (100-52-7); Biphenyl (92-52-4); ε-Caprolactam (105-60-2); Chlorobenzilate (510-15-6); 1-Chloronaphthalene (90-13-1); Diallate (2303-16-4); Dibenz(a,j)acridine (224-42-0); 2,6-Dichlorophenol (87-65-0); 7,12-Dimethylbenz(a)anthracene (57-97-6); 1,4-Dioxane (123-91-1); Diphenyl ether (101-84-8); Ethyl methacrylate (97-63-2); Ethyl methanesulfonate (62-50-0); Hexachloropropene (1888-71-7); Isodrin (465-73-6); Isosafrole (cis & trans) (120-58-1); Kepone (143-50-0); 3-Methylcholanthrene (56-49-5); Methyl methanesulfonate (66-27-3); 1,4-Naphthoquinone (130-15-4); 4-Nitroquinoline-N-oxide (56-57-5); Pentachlorobenzene (608-93-5); Pentachloroethane (76-01-7); Pentachloronitrobenzene (quintozene) (82-68-8); Phenacetin (62-44-2); Propyzamide (23950-58-5); Safrole (94-59-7); 1,2,4,5-Tetrachlorobenzene (95-94-3); 1,3,5-Trinitrobenzene (99-35-4)
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty