Would you like to log in again?
Antifoam Agent for Purge-and-Trap Samples, Restek
Supplier: Restek
31822
RK31822EA 49.47 USD InStock
RK31822
Antifoam Agent for Purge-and-Trap Samples, Restek
Standards Organic Standards
Foam generated as purge gas passes through a sample can enter the analytical trap and possibly the GC column. This silica-containing antifoam agent is effective over a wide pH range and will not conflict with chromatography of target analytes.
  • Efficiently controls foam; effective over a wide pH range.
  • No hazardous materials, no components that are target analytes.
  • Effective at less than 0.1% of sample volume.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty