Would you like to log in again?
CLP–Pesticides Toxaphene Standard, Restek
Supplier: Restek
32005
RK32005EA 50.44 USD InStock
RK32005
CLP–Pesticides Toxaphene Standard, Restek
Standards Organic Standards
Standard is packaged in hexane at a concentration of 1000 µg/mL in a 1 mL ampoule.
  • Toxaphene standards may exhibit lot-to-lot variation
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty