Would you like to log in again?
Revised GPC Calibration Mix (5 components), Restek
Supplier: Restek
32041 32042
RK32041EA 32.01 USD InStock
RK32041 RK32042
Revised GPC Calibration Mix (5 components), Restek
Standards Organic Standards
Contains: Bis(2-ethylhexyl) phthalate (117-81-7), 5 mg/mL; Corn oil (8001-30-7), 250 mg/mL; Methoxychlor (72-43-5), 1.0 mg/mL; Perylene (198-55-0), 0.2 mg/mL; Sulfur (7704-34-9), 0.8 mg/mL
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty