Would you like to log in again?
608 Complete Kit, Restek
Supplier: Restek
32060
RK32060EA 358.9 USD InStock
RK32060
608 Complete Kit, Restek
Standards Organic Standards
This kit contains 1 ml of each of these mixtures 608 calibration mix, Aroclor 1016, Aroclor 1221, Aroclor 1232, Aroclor 1242, Aroclor 1248, Aroclor 1254, Aroclor 1260, toxaphene, chlordane.
  • Aroclor, toxaphene, and chlordane standards may exhibit lot-to-lot variation.
Contains: 32022: 608 Calibration Mix; Aldrin (309-00-2); α-BHC (319-84-6); β-BHC (319-85-7); δ-BHC (319-86-8); γ-BHC (Lindane) (58-89-9); 4,4'-DDD (72-54-8); 4,4'-DDE (72-55-9); 4,4'-DDT (50-29-3); Dieldrin (60-57-1); Endosulfan I (959-98-8); Endosulfan II (33213-65-9); Endosulfan sulfate (1031-07-8); Endrin (72-20-8); Endrin aldehyde (7421-93-4); Heptachlor (76-44-8); Heptachlor epoxide (isomer B) (1024-57-3);32006: Aroclor 1016; Aroclor 1016;32007: Aroclor 1221; Aroclor 1221;32008: Aroclor 1232; Aroclor 1232;32009: Aroclor 1242; Aroclor 1242;32010: Aroclor 1248; Aroclor 1248;32011: Aroclor 1254; Aroclor 1254;32012: Aroclor 1260; Aroclor 1260;32005: Toxaphene Solutions; Toxaphene (8001-35-2);32021: Chlordane; chlordane
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty