Would you like to log in again?
Herbicide Lab Performance Check Mix (5 components), Restek
Supplier: Restek
32063
RK32063EA 43.65 USD InStock
RK32063
Herbicide Lab Performance Check Mix (5 components), Restek
Standards Organic Standards
This mix contains five components.
Components are dinoseb methyl ether (4 μg/ml), DCAA methyl ester (500 μg/ml), 4,4'-dibromooctafluorobiphenyl (250 μg/ml), 3,5-dichlorobenzoic acid methyl ester (600 µg/ml), 4-nitroanisole (1600 μg/ml).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty