Would you like to log in again?
531.2 Carbamate Pesticide Calibration Mixture (11 components), Restek
Supplier: Restek
32435
RK32435EA 62.08 USD InStock
RK32435
531.2 Carbamate Pesticide Calibration Mixture (11 components), Restek
Standards Organic Standards
This mix contains 11 components.
  • Complete set of materials for N-methylcarbamoyloximes and N-methylcarbamates.
  • New mix satisfies latest update of EPA Method 531.2.
  • Formulated in acetonitrile for stability and convenience for LC analysis.
Because carbamates, especially aldicarb sulfone, are unstable, carbamates analyses usually are HPLC based. The U.S. EPA method for monitoring these compounds in drinking water, Method 531, calls for HPLC with post-column derivatization. 1-Naphthol, a fluorescent metabolite of carbaryl, was added to the analyte list in 2003. Our 531.2 Carbamate Pesticides Calibration Mixture includes 1-naphthol. For stability, we formulate the mix in acetonitrile.
Contains: Aldicarb (116-06-3); Aldicarb sulfone (1646-88-4); Aldicarb sulfoxide (1646-87-3); Carbaryl (Sevin) (63-25-2); Carbofuran (1563-66-2); 3-Hydroxycarbofuran (16655-82-6); Methiocarb (2032-65-7); Methomyl (16752-77-5); 1-Naphthol (90-15-3); Oxamyl (23135-22-0); Propoxur (Baygon) (114-26-1).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty