Would you like to log in again?
FAME #5 Mix (5 components), Restek
Supplier: Restek
35014
RK35014EA 51.2 USD InStock
RK35014
FAME #5 Mix (5 components), Restek
Standards Organic Standards
Contains: Chain, Compound (CAS#), % by Weight; C16:01 Methyl palmitoleate (1120-25-8), 20%; C18:01 Methyl oleate (112-62-9), 20%; C20:01 Methyl eicosenoate (20839-34-3), 20%; C22:01 Methyl erucate (1120-34-9), 20%; C24:01:00 Methyl nervonate (2733-88-2), 20%
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty