Would you like to log in again?
FAME #6 Mix (5 components), Restek
Supplier: Restek
35015
RK35015EA 51.2 USD InStock
RK35015
FAME #6 Mix (5 components), Restek
Standards Organic Standards
This mixture can be used for quantification (AOCS Method CE 1a–13) and approximate the compositions of the following types of oils: FAME #6:oils of long chain lengths (C20 - C21)
Contains: Chain, Compound (CAS#), % by Weight; C20:00 Methyl arachidate (1120-28-1), 20%; C20:01 Methyl eicosenoate (20839-34-3), 20%; C20:02 Methyl eicosadienoate (2463-02-7), 20%; C20:03 Methyl homo gamma linolenate (16326-32-2), 20%; C20:04 Methyl arachidonate (2566-89-4), 20%
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty