Would you like to log in again?
MXT®-502.2 Columns (Siltek®-treated stainless steel), Restek
Supplier: Restek
70908 70910 70919
10849-034EA 1061.03 USD InStock
10849-034 10851-058 RK70910
MXT®-502.2 Columns (Siltek®-treated stainless steel), Restek
Chromatography Columns
An MXT®-502.2 column will enable you to quantify all compounds listed in U.S.
  • Application-specific columns with unique selectivity for volatile organic pollutants
  • Excellent separation of trihalomethanes; ideal polarity for light hydrocarbons and aromatics
  • Temperature range: −20 to 320 °C
  • 4.5" standard coil diameter
  • Proprietary Crossbond® diphenyl/dimethyl polysiloxane phase
The diphenyl/dimethyl polysiloxane based MXT®-502.2 stationary phase provides low bleed and thermal stability to 320 °C. A 105 m column can separate the light gases specified in EPA methods without subambient cooling.
EPA methods 502.2 or 524.2, whether you use a mass spectrometer or a PID in tandem with an ELCD.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty