Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DIABLO Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-244 100-401-244
RL-100-401-244EA 518 USD InStock
CA100-401-244 RL-100-401-244
Anti-DIABLO Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with human Smac/DIABLO protein by immunoblot.  Lysates from human HeLa and LNCaP cells are positive for Smac/DIABLO.  Other animal tissues have not been tested.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:1000 - 1:2000; Immunoprecipitation: 1:100; Other Dilution: User Optimized

Type: Primary
Antigen: DIABLO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty