Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ANGPT1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-403 100-401-403
RL-100-401-403EA 518 USD InStock
RL-100-401-403 CA100-401-403
Anti-ANGPT1 Rabbit Polyclonal Antibody
Antibodies
Suitable for western blotting, Immunohistochemistry (IHC) and other antibody based assays.  A 1:500 dilution is recommended for western blotting.  The reaction of this antiserum directly with cell supernatants may result in high background due to reactivi

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500-1:2000; Immunochemistry: 1:200-1:800; Other Dilution: User Optimized

Type: Primary
Antigen: ANGPT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty