Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MBP Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-385 200-401-385
RL-200-401-385EA 575 USD InStock
RL-200-401-385 CA200-401-385
Anti-MBP Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-MALTOSE BINDING PROTEIN (MBP) EPITOPE TAG Reacts with Pan

Type: Primary
Antigen: MBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: pan

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty