Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GST Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
600-106-200 600-106-200
RL-600-106-200EA 771 USD InStock
CA600-106-200 RL-600-106-200
Anti-GST Goat Polyclonal Antibody (Biotin)
Antibodies
Primary Goat Anti-GLUTATHIONE-S-TRANSFERASE (Schistosoma japonicum) IgG Fraction Reacts with Pan

Type: Primary
Antigen: Glutathion-S-Transferase
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Pan

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty