Would you like to log in again?
Chicken IgG (AP (Alkaline Phosphatase))
Supplier: Rockland Immunochemicals
003-0502
RL003-0502EA 244 USD InStock
RL003-0502
Chicken IgG (AP (Alkaline Phosphatase))
Antibodies
Chicken IgG whole molecule Alkaline Phosphatase
  • control or standard reagent
Secreted as part of the adaptive immune response by plasma B cells, immunoglobulin G constitutes 75% of serum immunoglobulins. Immunoglobulin G binds to viruses, bacteria, as well as fungi and facilitates their destruction or neutralization via agglutination (and thereby immobilizing them), activation of the compliment cascade, and opsinization for phagocytosis.
The whole IgG molecule possesses both the F(c) region, recognized by high-affinity Fc receptor proteins, as well as the F(ab) region possessing the epitope-recognition site.
Both heavy and light chains of the antibody molecule are present.
Chicken IgG whole molecule is ideal for investigators in Cancer, Immunology, and Microbiology research.
Chicken IgG whole molecule alkaline phosphatase can be utilized as a control or standard reagent in Western Blotting and ELISA experiments.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty