Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG2c kappa Rat Monoclonal Antibody [clone: RG2cK]

Supplier: Rockland Immunochemicals
012-001-P04 012-001-P04
RL012-001-P04EA 252 USD InStock
RL012-001-P04 CA012-001-P04
Anti-IgG2c kappa Rat Monoclonal Antibody [clone: RG2cK]
Antibodies
Rat IgG2c Kappa isotype control can be utilized as a control or standard reagent in Western Blotting, Flow Cytometry, and ELISA experiments where determination of sample isotype is important. Specific conditions should be optimized by user.

Recommended Dilutions: Flow Cytometry: 1:1000-1:5000;

Type: Control
Antigen: IgG2c kappa
Clonality: Monoclonal
Clone: RG2cK
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2c kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty