Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HRP Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-1138 100-1138
RL100-1138EA 309 USD InStock
CA100-1138 RL100-1138
Anti-HRP Goat Polyclonal Antibody
Antibodies
Anti-Peroxidase (Horesradish) (Goat) Antibody has been assayed against 1.0 µg of Peroxidase (Horseradish) in a standard ELISA using Alkaline Phosphatase conjugated Affinity Purified anti-Goat IgG (H&L) (Rabbit) and pNPP as a substrate.

Recommended Dilutions: ELISA: 1:9000 - 1:40000; Western Blot: 1:900 - 1:4000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Tag
Antigen: HRP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: pan

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty