Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PFKM Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-1156 100-1156
RL100-1156EA 1290 USD InStock
RL100-1156 CA100-1156
Anti-PFKM Goat Polyclonal Antibody
Antibodies
This purified antibody has been tested for use in ELISA, immunofluorescence microscopy and western blot.  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 48 kDa in size.

Recommended Dilutions: ELISA: 1:4000 - 1:20000; Western Blot: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: PFKM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: pan

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty