Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EP300 Mouse Polyclonal Antibody [clone: AC240]

Supplier: Rockland Immunochemicals
100-301-176 100-301-176
RL100-301-176EA 528 USD InStock
CA100-301-176 RL100-301-176
Anti-EP300 Mouse Polyclonal Antibody [clone: AC240]
Antibodies
This product was assayed by immunoblot and found to be reactive against p300 at a dilution of 1:10 to 1:50 followed by reaction with Peroxidase conjugated Affinity Purified anti-Mouse IgG (H&L) (Goat).

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000;

Type: Primary
Antigen: EP300
Clonality: Polyclonal
Clone: AC240
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty