Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CPB1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-049 100-401-049
RL100-401-049EA 1290 USD InStock
RL100-401-049 CA100-401-049
Anti-CPB1 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-CARBOXYPEPTIDASE B (Porcine Pancreas) Reacts with Pan

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: CPB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty