Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NFYA Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-100 100-401-100
RL100-401-100EA 518 USD InStock
CA100-401-100 RL100-401-100
Anti-NFYA Rabbit Polyclonal Antibody
Antibodies
Anti-NF-Y (A subunit) Antibody was assayed by immunoblot and found to be reactive against NF-Y (A subunit specific) at a dilution of 1:6,000 followed by reaction with Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat).

Recommended Dilutions: ELISA: 1:20000 - 1:100000; GelShift: Use 2.0 µl; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:400;

Type: Primary
Antigen: NFYA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty