Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EGFR Rabbit Monoclonal Antibody [clone: 15A3]

Supplier: Rockland Immunochemicals
100-401-149 100-401-149
RL100-401-149EA 518 USD InStock
RL100-401-149 CA100-401-149
Anti-EGFR Rabbit Monoclonal Antibody [clone: 15A3]
Antibodies
Anti-EGFR antibody is specifically designed for ELISA, immunoblotting and immunoprecipitation.  Reactivity in other assays is likely, but has not been determined.  Recognition of EGFR is independent of the phosphorylation status at tyrosine 1173.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 2.5 µg/ml; Immunoprecipitation: 10 µl;

Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: 15A3
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty