Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CCNT1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-158 100-401-158
RL100-401-158EA 530 USD InStock
RL100-401-158 CA100-401-158
Anti-CCNT1 Rabbit Polyclonal Antibody
Antibodies
Suitable for ELISA, immunoprecipitation, immunochemistry, immunoblotting and other immunological methods requiring high titer and specificity.  Anti-Cyclin T1 is suitable for the detection by immunoblot of human, Rat and mouse Cyclin T1.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:200 - 1:1000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CCNT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty