Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CDK4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-162 100-401-162
RL100-401-162EA 518 USD InStock
RL100-401-162 CA100-401-162
Anti-CDK4 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-cdk4 Reacts with Human, Mouse, Rat

Recommended Dilutions: ELISA: 1:500 - 1:2000; Western Blot: 1:200 - 1:500; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CDK4
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty