Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RBL2 Rabbit Monoclonal Antibody [clone: 12.A1.1]

Supplier: Rockland Immunochemicals
100-401-173 100-401-173
RL100-401-173EA 518 USD InStock
RL100-401-173 CA100-401-173
Anti-RBL2 Rabbit Monoclonal Antibody [clone: 12.A1.1]
Antibodies
Suitable for ELISA, immunoprecipitation, immunoblotting, immunochemistry, and other immunological methods requiring high titer and specificity.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:200 - 1:1000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: RBL2
Clonality: Monoclonal
Clone: 12.A1.1
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty