Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CREB1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-195 100-401-195
RL100-401-195EA 518 USD InStock
RL100-401-195 CA100-401-195
Anti-CREB1 Rabbit Polyclonal Antibody
Antibodies
Anti-CREB-1 Antibody is suitable for use in SDS, Western Blot, and ELISA assays. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Gel Shift: 0.5 to 1.0 µl per assay; Western Blot: 1:500 - 1:1000;

Type: Primary
Antigen: CREB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty