Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DAB1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-225S
RL100-401-225SEA 140 USD InStock
RL100-401-225S
Anti-DAB1 Rabbit Polyclonal Antibody
Antibodies
Anti-Dab1 antibody is suitable for the detection of Dab1 by immunoprecipitation.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:5000; Immunoprecipitation: 1 µl per 500 µg;

Type: Primary
Antigen: DAB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty