Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PTGS2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-226 100-401-226
RL100-401-226EA 468 USD InStock
RL100-401-226 CA100-401-226
Anti-PTGS2 Rabbit Polyclonal Antibody
Antibodies
Cox-2 (Cyclooxygenase-2) is an inducible enzyme that is normally absent from cells, however, in response to growth factors, tumor promoters and some cytokines, it undergoes a rapid and transient expression.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:5000 - 1:10000;

Type: Primary
Antigen: PTGS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty