Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SPP1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-404 100-401-404
RL100-401-404EA 504 USD InStock
RL100-401-404 CA100-401-404
Anti-SPP1 Rabbit Polyclonal Antibody
Antibodies
Anti-Osteopontin is suitable for western blotting, immunochemistry (formalin-fixed paraffin-embedded sections) and ELISA. The antibody exclusively recognizes C-terminal fragments of both thrombin and MMP-cleaved OPN.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:100 - 1:300; Immunoprecipitation: 1:100;

Type: Primary
Antigen: SPP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat, Pig

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty