Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-tetX Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-894 100-401-894
RL100-401-894EA 523 USD InStock
RL100-401-894 CA100-401-894
Anti-tetX Rabbit Polyclonal Antibody
Antibodies
This antibody has been tested for use in ELISA and western blot.  For western blots expect a band of approximately 52 kDa in size corresponding to full-length TTFC.  Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:50000 - 1:250000; Western Blot: 1:1000 - 1:40000;

Type: Primary
Antigen: tetX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Clostridium tetani

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty