Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CCNT2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-931 100-401-931
RL100-401-931EA 518 USD InStock
RL100-401-931 CA100-401-931
Anti-CCNT2 Rabbit Polyclonal Antibody
Antibodies
This antibody has been tested for use in ELISA and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect bands approximately 74 and 81 kDa in size corresponding to isoforms of Cyclin T2 Protein.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:2000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CCNT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty