Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CUL3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-A03 100-401-A03
RL100-401-A03EA 518 USD InStock
RL100-401-A03 CA100-401-A03
Anti-CUL3 Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with human Cul3 by western blot and immunoprecipitation.  The antibody immunoprecipitates in vitro translated product and Protein from cell lysates (using HeLa or NIH-3T3).  An 88.9 kDa band corresponding to human Cul3 is detected.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CUL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty