Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CUL5 Rabbit Monoclonal Antibody [clone: 14C5]

Supplier: Rockland Immunochemicals
100-401-A05 100-401-A05
RL100-401-A05EA 518 USD InStock
RL100-401-A05 CA100-401-A05
Anti-CUL5 Rabbit Monoclonal Antibody [clone: 14C5]
Antibodies
This antibody reacts with human Cul5 by western blot and immunoprecipitation.  The antibody immunoprecipitates Protein from cell lysates (using 293T, U2OS and others).  This antibody also co-immunoprecipitates associated Proteins.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000;

Type: Primary
Antigen: CUL5
Clonality: Monoclonal
Clone: 14C5
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty