Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CUL9 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-A07 100-401-A07
RL100-401-A07EA 518 USD InStock
RL100-401-A07 CA100-401-A07
Anti-CUL9 Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with human and mouse PARC by western blot and immunoprecipitation.  The antibody immunoprecipitates Protein from cell lysates (using HeLa, NIH-3T3, and others).  To date co-immuno-precipitation using 35S-IP has been negative.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000;

Type: Primary
Antigen: CUL9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty