Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RBX1 Rabbit Monoclonal Antibody [clone: 14A11]

Supplier: Rockland Immunochemicals
100-401-A13 100-401-A13
RL100-401-A13EA 518 USD InStock
RL100-401-A13 CA100-401-A13
Anti-RBX1 Rabbit Monoclonal Antibody [clone: 14A11]
Antibodies
This antibody reacts with human ROC1 by western blot and immunoprecipitation.  The antibody immunoprecipitates in vitro translated Protein and Protein from overexpressing cell lysates (using HeLa and NIH-3T3, and others).

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000;

Type: Primary
Antigen: RBX1
Clonality: Monoclonal
Clone: 14A11
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty