Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NSP8 Rabbit Polyclonal Antibody [clone: 13F9]

Supplier: Rockland Immunochemicals
100-401-A53 100-401-A53
RL100-401-A53EA 471 USD InStock
CA100-401-A53 RL100-401-A53
Anti-NSP8 Rabbit Polyclonal Antibody [clone: 13F9]
Antibodies
This antibody has been tested for use in immunofluorescence microscopy, immunoelectron microscopy, immunoprecipitation and by western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 22 kDa.

Recommended Dilutions: Immunofluorescence: 1:300; Western Blot: 1:1000; Immunoprecipitation: 1:60; Immunoelectron Microscopy 1:100

Type: Primary
Antigen: NSP8
Clonality: Polyclonal
Clone: 13F9
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: SARS Virus

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty