Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NSP13 Rabbit Polyclonal Antibody [clone: 5D11.G5]

Supplier: Rockland Immunochemicals
100-401-A54 100-401-A54
RL100-401-A54EA 458 USD InStock
CA100-401-A54 RL100-401-A54
Anti-NSP13 Rabbit Polyclonal Antibody [clone: 5D11.G5]
Antibodies
This antibody has been tested for use in western blotting, immunofluorescence microscopy and immunoelectron microscopy. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 67 kDa in size.

Recommended Dilutions: Immunofluorescence: 1:800; Western Blot: 1:1000; Immunoprecipitation: 1:60; Immunoelectron Microscopy 1:300

Type: Primary
Antigen: NSP13
Clonality: Polyclonal
Clone: 5D11.G5
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: SARS Virus

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty