Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CES1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-B14S 100-401-B14S
RL100-401-B14SEA 148 USD InStock
RL100-401-B14S CA100-401-B14S
Anti-CES1 Rabbit Polyclonal Antibody
Antibodies
This antibody has been tested for use in ELISA and western blotting and is specific for human CE1 (carboxylesterase).  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 66 kDa in size corresponding CE1 b.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:2500 – 1:5000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CES1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty