Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BAG6 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-B63 100-401-B63S
RL100-401-B63SEA 148 USD InStock
RL100-401-B63S RL100-401-B63
Anti-BAG6 Rabbit Polyclonal Antibody
Antibodies
This Scythe polyclonal antibody has been tested for use in western blotting and is specific for mouse Scythe/Bat3 and its isoforms. Specific conditions for reactivity should be optimized by the end user. Expect multiple bands.

Recommended Dilutions: Western Blot: 1:5000 to 1:8000

Type: Primary
Antigen: BAG6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Rat, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty