Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GRIN2A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
612-401-D89 100-401-C86
RL612-401-D89EA 518 USD InStock
RL612-401-D89 RL100-401-C86
Anti-GRIN2A Rabbit Polyclonal Antibody
Antibodies
Anti-NMDA R2A Antibody is suitable for use in Western Blotting and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 180 kDa in size corresponding to the NR2A subuint of the NMDA receptor.

Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:500;

Type: Primary
Antigen: GRIN2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty