Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SYN1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-C92 100-401-C92
RL100-401-C92EA 518 USD InStock
RL100-401-C92 RL600-401-C92
Anti-SYN1 Rabbit Polyclonal Antibody
Antibodies
Anti-Synapsin I antibody is suitable for Western Blotting, Immunoprecipitation, Immunochemistryand IF. Expect a band of approximately 78 kDa in size corresponding to the synapsin I doublet in Western Blots of Rat brain extracts.

Recommended Dilutions: Immunofluorescence: 1:2000; Western Blot: 1:1000; Immunochemistry: 1:2000; Immunoprecipitation: 1µg per 200µg lysate
0.05 M Tris Chloride, 0.15 M Sodium Chloride, pH 7.4;

Type: Primary
Antigen: SYN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty