Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CNP Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-D21
RL100-401-D21EA 518 USD InStock
RL100-401-D21
Anti-CNP Rabbit Polyclonal Antibody
Antibodies
Anti-CNP is suitable for usage in ELISA, Western Blotting and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a 46kDa band by western blot corresponding to CNP Protein in the appropriate cell lysate of tissue.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000; Immunochemistry: 1:1000

Type: Primary
Antigen: CNP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty