Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GSTK1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-D29
RL100-401-D29EA 518 USD InStock
RL100-401-D29
Anti-GSTK1 Rabbit Polyclonal Antibody
Antibodies
Anti-Dsba-L Antibody are suitable for usage in ELISA, Western Blotting and IHC. Specific conditions for reactivity should be optimized by the end user. It is suggested that you add 1% BSA to the antibody dilution buffer.

Recommended Dilutions: ELISA: 1:20.000; Western Blot: 1:2000; Immunochemistry: 1:200

Type: Primary
Antigen: Gstk1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty